Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slfn5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_183201.4 | NP_899024.3 | 5 | 1755 | ||
GenBank mRNA | AK036486.1 | - | 5 | 1756 | ||
AK040359.1 | - | 5 | 1742 | |||
AY261804.1 | - | 4 | 1717 | |||
AY261805.1 | - | 4 | 1712 | |||
BC117909.1 | - | 4 | 1635 | 40097236 | ||
BC117910.1 | - | 4 | 1635 | 40097237 |