Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rft1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177815.3 | NP_808483.2 | 5 | 511 | ||
GenBank mRNA | AK086390.1 | - | 5 | 500 | ||
BC082539.1 | - | 5 | 479 | |||
BC094336.1 | - | 5 | 479 | |||
BC116368.2 | - | 5 | 492 | |||
BC116369.2 | - | 5 | 492 | |||
BC141275.1 | - | 5 | 505 | 9055887 | ||
BC145606.1 | - | 5 | 505 | 9054172 |