Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 9330182O14Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256056.1 | NP_001242985.1 | 3 | 472 | ||
GenBank mRNA | AK034361.1 | - | 3 | 474 |