Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Syt14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301370.1 | NP_001288299.1 | 5 | 704 | ||
NM_181546.3 | NP_853524.1 | 5 | 704 | |||
GenBank mRNA | AB102947.1 | - | 5 | 627 | ||
AK045901.1 | - | 5 | 705 | |||
AK051175.1 | - | 3 | 192 | |||
BC141262.1 | - | 5 | 646 | 9055874 | ||
BC145602.1 | - | 5 | 646 | 9054168 |