Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 4932418E24Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177841.3 | NP_808509.1 | 6, 7 | 2246 | ||
GenBank mRNA | AK077032.1 | - | 6, 7 | 2247 |