Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nostrin | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181547.3 | NP_853525.3 | 15, 16 | 1485 | ||
GenBank mRNA | AJ564264.1 | - | 15, 16 | 1395 | ||
AY280963.1 | - | 15, 16 | 1438 | |||
BC069942.1 | - | 15, 16 | 1462 | 30443942 |