Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dennd2c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177857.1 | NP_808525.2 | 17 | 2873 | ||
GenBank mRNA | AK137637.1 | - | 12 | 1480 | ||
AK172232.1 | - | 13 | 1595 | |||
BC046440.1 | - | 13 | 1341 | |||
BC060666.1 | - | 17 | 2873 | 6841982 |