Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dennd2c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177857.1 | NP_808525.2 | 2 | 1044 | ||
GenBank mRNA | AK044389.1 | - | 2 | 1027 | ||
BC060666.1 | - | 2 | 1044 | 6841982 |