Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zc3h4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198631.2 | NP_941033.2 | 3 | 400 | ||
GenBank mRNA | AK138078.1 | - | 2 | 2218 | ||
AK151148.1 | - | 4 | 452 | |||
AK151306.1 | - | 4 | 452 | |||
BC057627.1 | - | 3 | 345 | 6412890 | ||
BY167917.1 | - | 4 | 449 | |||
CJ101946.1 | - | 3 | 405 |