Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Saxo2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177894.4 | NP_808562.3 | 3 | 815 | ||
GenBank mRNA | AK051095.1 | - | 3 | 811 | ||
BC144779.1 | - | 3 | 760 | |||
BC144780.1 | - | 3 | 777 |