Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Saxo2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177894.4 | NP_808562.3 | 2 | 291 | ||
GenBank mRNA | AK038727.1 | - | 3 | 471 | ||
AK051095.1 | - | 2 | 287 |