Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dock1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033420.2 | NP_001028592.1 | 16 | 1630 | ||
GenBank mRNA | AK138143.1 | - | 16 | 1577 | ||
AK147836.1 | - | 16 | 1628 | |||
AK164963.1 | - | 16 | 1615 |