Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Piwil4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177905.3 | NP_808573.2 | 18 | 2479 | ||
GenBank mRNA | AB258534.1 | - | 19 | 2563 | ||
AK033746.1 | - | 403 | ||||
AK143022.1 | - | 10 | 1378 |