Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Serpina7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177920.5 | NP_808588.3 | 5 | 1150 | ||
GenBank mRNA | AK050361.1 | - | 6 | 695 | ||
AK149459.1 | - | 5 | 1131 | |||
AY575783.1 | - | 4 | 1084 | |||
BC132100.1 | - | 4 | 1145 | 40130377 |