Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Krt78 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_212487.4 | NP_997652.4 | 9 | 1455 | ||
GenBank mRNA | AK146611.1 | - | 9 | 1452 | ||
BC119600.1 | - | 9 | 1392 | |||
BC119601.1 | - | 9 | 1392 | |||
BC127241.1 | - | 9 | 1392 | |||
BC127618.1 | - | 9 | 1392 |