Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zscan10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033425.4 | NP_001028597.2 | 6 | 1228 | ||
NM_001289482.1 | NP_001276411.1 | 4 | 898 | |||
GenBank mRNA | AK141259.1 | - | 6 | 1199 | ||
BC068284.1 | - | 5 | 901 | |||
DQ323929.1 | - | 6 | 1196 | |||
DQ323931.1 | - | 4 | 866 | |||
EF152498.1 | - | 5 | 1043 | |||
EF152502.1 | - | 4 | 820 | |||
EF152503.1 | - | 2 | 697 |