Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lyg2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033427.3 | NP_001028599.1 | 4, 5 | 666 | ||
GenBank mRNA | AK132442.1 | - | 4, 5 | 668 | ||
BC147562.1 | - | 4, 5 | 603 | 9056552 | ||
BC147567.1 | - | 4, 5 | 603 | 9056557 |