Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp36l3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009549.2 | NP_001009549.1 | 1 | 547 | ||
GenBank mRNA | AY661338.1 | - | 1 | 547 |