Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem220 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291042.1 | NP_001277971.1 | 6 | 411 | ||
NM_001291043.1 | NP_001277972.1 | 6 | 407 | |||
NM_177392.3 | NP_796366.1 | 7 | 483 | |||
GenBank mRNA | AK043094.1 | - | 7 | 484 | ||
AK078505.1 | - | 6 | 410 | |||
BC110628.1 | - | 6 | 411 | 40052084 | ||
BC110629.1 | - | 6 | 407 | 40052085 |