Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pramel6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178249.2 | NP_839980.1 | 3 | 991 | ||
GenBank mRNA | AF490342.1 | - | 3 | 991 | ||
AK139830.1 | - | 4 | 1225 |