Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc2a12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178934.4 | NP_849265.2 | 4 | 1789 | ||
GenBank mRNA | AJ549317.2 | - | 4 | 1789 | ||
AK028970.1 | - | 4 | 1762 | |||
AK031659.1 | - | 4 | 1707 | |||
AK035972.1 | - | 4 | 1670 | |||
AK143263.1 | - | 4 | 1778 | |||
AY230881.1 | - | 4 | 1688 | |||
BC096454.1 | - | 4 | 1720 | 4223231 | ||
BC116314.1 | - | 4 | 1650 | 40095492 | ||
BC116315.1 | - | 4 | 1653 | 40095493 | ||
BC138316.1 | - | 4 | 1636 | 8861336 | ||
BC138317.1 | - | 4 | 1636 | 8861337 |