Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trim46 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039466.1 | NP_001034555.1 | 8 | 1343 | ||
NM_183037.2 | NP_898858.1 | 8 | 1446 | |||
GenBank mRNA | AK132368.1 | - | 8 | 1313 | ||
AK170904.1 | - | 8 | 1344 | |||
AY251388.1 | - | 8 | 1326 | |||
BC065049.1 | - | 9 | 1527 | 6418851 |