Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Col27a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025685.3 | NP_079961.3 | 3 | 1928 | ||
GenBank mRNA | AK157951.1 | - | 3 | 1929 | ||
AY167568.2 | - | 3 | 1514 |