Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tlcd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291156.1 | NP_001278085.1 | 4 | 549 | ||
NM_027249.3 | NP_081525.1 | 4 | 549 | |||
GenBank mRNA | AK008522.1 | - | 4 | 525 | ||
BC021945.1 | - | 4 | 520 | 5098319 | ||
CV562896.1 | - | 4 | 550 |