Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tbc1d22b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198647.1 | NP_941049.1 | 4, 5 | 731 | ||
GenBank mRNA | BC045600.1 | - | 4, 5 | 731 | 4489206 |