Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Iars2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198653.2 | NP_941055.1 | 15 | 2011 | ||
GenBank mRNA | AK036182.1 | - | 15 | 1979 | ||
AK043590.1 | - | 15 | 2013 | |||
AK048653.1 | - | 15 | 2012 | |||
AK050358.1 | - | 15 | 1994 | |||
AK051778.1 | - | 15 | 1972 | |||
AK078183.1 | - | 15 | 2013 | |||
AK085714.1 | - | 15 | 2009 | |||
AK088965.1 | - | 15 | 1972 | |||
AK166832.1 | - | 15 | 1994 | |||
BC027432.1 | - | 5 | 469 | |||
BC037123.1 | - | 10 | 1071 | |||
BC052403.1 | - | 15 | 1988 | 5715723 |