Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpr176 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_201367.3 | NP_958755.1 | 3 | 940 | ||
GenBank mRNA | AY255554.1 | - | 2 | 459 | ||
BC052037.1 | - | 3 | 832 | 5702597 |
Set Membership: |
G-protein coupled receptor |