Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc4a5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166067.1 | NP_001159539.1 | 7 | 1072 | ||
GenBank mRNA | CO431509.1 | - | 2 | 122 | ||
CX562762.1 | - | 4 | 384 |