Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Glipr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027450.3 | NP_081726.1 | 4, 5 | 370 | ||
GenBank mRNA | AK017557.1 | - | 4, 5 | 347 | ||
AK089960.1 | - | 4, 5 | 350 | |||
AK141459.1 | - | 4, 5 | 359 | |||
AK154433.1 | - | 4, 5 | 348 | |||
AK167595.1 | - | 4, 5 | 370 | |||
BC031750.1 | - | 4, 5 | 370 | 4037002 |