Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hcrtr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198962.3 | NP_945200.1 | 4 | 807 | ||
GenBank mRNA | AF394597.1 | - | 3 | 436 | ||
AK038551.1 | - | 4 | 808 | |||
AK048781.1 | - | 4 | 807 | |||
AK079572.1 | - | 4 | 840 | |||
AY336084.1 | - | 4 | 775 | |||
AY336085.1 | - | 4 | 775 | |||
BC140957.1 | - | 4 | 803 | 9055569 |
Set Membership: |
G-protein coupled receptor |