Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ugt1a5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_201643.2 | NP_964005.2 | 1 | 510 | ||
GenBank mRNA | AB094481.1 | - | 1 | 489 | ||
AY227196.1 | - | 1 | 510 | |||
BC139093.1 | - | 1 | 490 | 8862115 | ||
BC139094.1 | - | 1 | 490 | 8862116 |