Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc87 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207268.3 | NP_997151.2 | 1 | 2275 | ||
GenBank mRNA | AK029877.1 | - | 1 | 2227 | ||
BC145660.1 | - | 1 | 2180 | 40130963 |