Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tcaf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_203396.1 | NP_981933.1 | 2 | 1063 | ||
GenBank mRNA | AY528666.1 | - | 2 | 1063 |