Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bin2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270537.1 | NP_001257466.1 | 7 | 594 | ||
GenBank mRNA | BY751384.1 | - | 7 | 597 | ||
CN691011.1 | - | 3 | 245 |