Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 4933401B06Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK016598.1 | - | 1 | 681 |
Gene Symbol: Actr5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK188146.1 | - | 339 |
Gene Symbol: Gle1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028923.3 | NP_083199.1 | 11 | 1707 | ||
GenBank mRNA | AK016671.1 | - | 11 | 1689 | ||
AK028183.1 | - | 11 | 1691 | |||
AK133888.1 | - | 11 | 1659 | |||
AK145748.1 | - | 11 | 1683 | |||
AK161015.1 | - | 11 | 1653 | |||
AK161659.1 | - | 11 | 1690 | |||
AK207245.1 | - | 1 | 121 | |||
AK216189.1 | - | 1 | 121 | |||
BC026797.1 | - | 11 | 1687 | 3990668 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AK139674.1 | - | 554 | Chr. - - [-] on Build |