Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tet2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040400.2 | NP_001035490.2 | 1158 | |||
GenBank mRNA | DQ079067.1 | - | 3 | 1136 | ||
GU079949.1 | - | 1 | 776 |