Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gm12429 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277167.1 | NP_001264096.1 | 4 | 1195 |
Gene Symbol: Gm13298 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001085530.1 | NP_001078999.1 | 4 | 1195 |