Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gm42035 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | FJ654104.1 | - | 3609 | |||
FJ654105.1 | - | 3166 | ||||
FJ654106.1 | - | 3609 | ||||
FJ654107.1 | - | 3608 | ||||
FJ654108.1 | - | 3608 | ||||
FJ654112.1 | - | 3109 | ||||
FJ654131.1 | - | 3608 | ||||
FJ654132.1 | - | 3609 |
Gene Symbol: Gm3579 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AY140896.1 | - | 1 | 3608 | ||
BC066867.1 | - | 1 | 1516 | 30462835 |