Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gm42035 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK133569.1 | - | 1421 | |||
FJ654104.1 | - | 3169 | ||||
FJ654106.1 | - | 3169 | ||||
FJ654107.1 | - | 3169 | ||||
FJ654108.1 | - | 3169 | ||||
FJ654112.1 | - | 2669 | ||||
FJ654131.1 | - | 3168 | ||||
FJ654132.1 | - | 3169 |
Gene Symbol: Gm3579 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK158167.1 | - | 1 | 3311 | ||
AY140896.1 | - | 1 | 3168 | |||
BC066867.1 | - | 1 | 1076 | 30462835 |
Gene Symbol: LOC672025 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK156270.1 | - | 1 | 250 |