Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Whamm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004185.3 | NP_001004185.3 | 7 | 1534 | ||
GenBank mRNA | AK140311.1 | - | 7 | 1544 | ||
AK162084.1 | - | 9 | 1539 | |||
AK220273.1 | - | 7 | 1534 | |||
BC042749.2 | - | 8 | 1369 | 5322109 | ||
BC050804.1 | - | 7 | 997 | 6771347 |