Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nlrc5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033207.3 | NP_001028379.2 | 38 | 4790 | ||
GenBank mRNA | FJ889356.1 | - | 38 | 4790 |