Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmprss11c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001030297.2 | NP_001025468.1 | 8, 9 | 1044 | ||
GenBank mRNA | AM260217.1 | - | 8, 9 | 1044 | ||
BC147157.1 | - | 8, 9 | 1025 | 9056205 | ||
BC147158.1 | - | 8, 9 | 1025 | 9056165 |