Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024707.2 | NP_001019878.1 | 8 | 2755 | ||
GenBank mRNA | AK149314.1 | - | 7 | 2280 | ||
BC094349.1 | - | 2 | 849 |