Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024707.2 | NP_001019878.1 | 7 | 2098 | ||
GenBank mRNA | AK149314.1 | - | 6 | 1623 | ||
AK149337.1 | - | 2094 | ||||
BC094349.1 | - | 1 | 192 |