Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rgs3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134257.3 | NP_599018.3 | 11 | 1726 | ||
GenBank mRNA | AF350047.1 | - | 12 | 1601 | ||
AK004648.1 | - | 5 | 895 | |||
AK041392.1 | - | 5 | 903 | |||
BC033449.1 | - | 2 | 742 | 3599345 |