Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccin | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001002787.2 | NP_001002787.1 | 1 | 1071 | ||
GenBank mRNA | AK030197.1 | - | 1 | 1074 | ||
BC150949.1 | - | 1 | 1058 | 9087860 | ||
BC150952.1 | - | 1 | 1058 | 9087863 |