Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Armcx5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009575.5 | NP_001009575.2 | 4 | 2021 | ||
GenBank mRNA | AK133950.1 | - | 4 | 2023 | ||
AK166624.1 | - | 5 | 1897 | |||
BC027106.1 | - | 1 | 1054 | 5358405 |