Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: E2f6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033270.2 | NP_150373.2 | 5 | 810 | ||
GenBank mRNA | AB221552.1 | - | 5 | 571 | ||
AF032131.1 | - | 5 | 788 | |||
AK146792.1 | - | 6 | 872 | |||
AK149061.1 | - | 966 | ||||
AK183289.1 | - | 2 | 160 | |||
BC019166.1 | - | 6 | 872 | 4236246 | ||
BC037656.1 | - | 6 | 753 | 5358554 | ||
BC057929.1 | - | 5 | 798 | 5289475 |