Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hspa14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015765.2 | NP_056580.2 | 5 | 419 | ||
GenBank mRNA | AK031489.1 | - | 358 | |||
AK145937.1 | - | 5 | 373 | |||
AK153397.1 | - | 5 | 381 | |||
BC002056.1 | - | 5 | 347 | 3493119 | ||
BU505607.1 | - | 2 | 70 | |||
U08215.1 | - | 5 | 379 |