Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hspa14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015765.2 | NP_056580.2 | 11 | 1156 | ||
GenBank mRNA | AK031489.1 | - | 1095 | |||
AK145937.1 | - | 11 | 1110 | |||
AK153397.1 | - | 11 | 1118 | |||
BC002056.1 | - | 11 | 1084 | 3493119 | ||
BI734146.1 | - | 3 | 259 | |||
U08215.1 | - | 11 | 1122 |